Learn More
Sterlitech Corporation CF042 Cell Assembly, Crossflow, Hastelloy (CF042H)
SDP
Catalog No. 501952349
Encompass_Preferred
The Sterlitech CF042 membrane cell is a lab scale cross flow filtration unit designed to provide fast and accurate performance data with minimal amounts of product, expense, and time.
CF042 Membrane Cell Product Availability:
  • Cross flow cell units available in Delrin, PTFE, Stainless Steel, Hastelloy™, or Acrylic 
  • 1000 psi (69 bar) max operating pressure (PTFE and Acrylic versions are 400 psi)
  • Active membrane area: 42cm2
  • Please note: A feed flow pump is required to operate the CF042 cross flow cell unit. We recommend the Hydracell pump by Wanner Engineering, which is a positive displacement, diaphragm pump designed for high pressure, lower flow applications. Min power available is 110V, 60 Hz. 220 V and 50 Hz motors are also available. There may be an additional cost if a variable speed drive is required.
    Catalog No. 50-195-2349 Supplier Sterlitech Corporation Supplier No. 1160266
    Add to Cart
    Add to Cart
    Request a Quote
    The Sterlitech CF042 membrane cell is a lab scale cross flow filtration unit designed to provide fast and accurate performance data with minimal amounts of product, expense, and time.
    CF042 Membrane Cell Product Availability:
  • Cross flow cell units available in Delrin, PTFE, Stainless Steel, Hastelloy™, or Acrylic 
  • 1000 psi (69 bar) max operating pressure (PTFE and Acrylic versions are 400 psi)
  • Active membrane area: 42cm2
  • Please note: A feed flow pump is required to operate the CF042 cross flow cell unit. We recommend the Hydracell pump by Wanner Engineering, which is a positive displacement, diaphragm pump designed for high pressure, lower flow applications. Min power available is 110V, 60 Hz. 220 V and 50 Hz motors are also available. There may be an additional cost if a variable speed drive is required.