Learn More
NANOPARTZ INC CF11 GOLD NANOPARTICLES, 1ML
SDP
Catalog No. NC2219533
Encompass
CF11 Fluorophore Labeled Spherical Gold Nanoparticles; a 150nm diameter sphere, Cy5 fluorophore, with a Carboxyl polymer functionalization, in TRIS, at 50 OD-mLs (2.5mg/mL, OD=50, 1mL
This item is not returnable. View return policy
Catalog No. NC2219533 Supplier NANOPARTZ INC Supplier No. CF11150CY5FCTRIS50
CF11 Fluorophore Labeled Spherical Gold Nanoparticles; a 150nm diameter sphere, Cy5 fluorophore, with a Carboxyl polymer functionalization, in TRIS, at 50 OD-mLs (2.5mg/mL, OD=50, 1mL