Learn More
Julabo USA Inc Each, CF30 refrigerated/heating circulator -30…+150 °C
SDP
Catalog No. 507533890
Encompass_Preferred
Bath/Circulators, Julabo, CF30 Compact Refrigerated and Heating Circulator -30...+150 °C. Economy CF30 model is ideal for routine applications with a working temperature range from -30 to +150 °C with a pressure pump. Units feature a splash-proof keypad with LED temperature display, PID temperature control and RS232 interface for PC connection. Included with each unit: 2 each barbed fittings for tubing 8 and 12 mm inner dia. (pump connections M16x1 female).
Catalog No. 50-753-3890 Supplier Julabo USA Inc Supplier No. 9400330.2
Add to Cart
Add to Cart
Request a Quote
Bath/Circulators, Julabo, CF30 Compact Refrigerated and Heating Circulator -30...+150 °C. Economy CF30 model is ideal for routine applications with a working temperature range from -30 to +150 °C with a pressure pump. Units feature a splash-proof keypad with LED temperature display, PID temperature control and RS232 interface for PC connection. Included with each unit: 2 each barbed fittings for tubing 8 and 12 mm inner dia. (pump connections M16x1 female).