Learn More
Julabo USA Inc Each, CF31 refrigerated/heating circulator -30…+200 °C
SDP
Catalog No. 507533892
Encompass_Preferred
Bath/Circulators, Julabo, HighTech CF31 unit has a working temperature range from -30 to +200 °C and includes a dual screen display with numeric keypad for easy data entry and instrument reading. RS232 port for external communication and Pt100 sensor connection for external monitoring and control. A 4-speed centrifugal pressure / suction pump provides 22-26 L/min flow with 5.8-10.2 psi pressure / 2.9-5.6 psi suction. Integrated programmer with 6 x 60 program steps. Included with each unit: 2 each barbed fittings for tubing 8 and 12 mm inner dia. (pump connections M16x1 female).
Catalog No. 50-753-3892 Supplier Julabo USA Inc Supplier No. 9400331.2
Add to Cart
Add to Cart
Request a Quote
Bath/Circulators, Julabo, HighTech CF31 unit has a working temperature range from -30 to +200 °C and includes a dual screen display with numeric keypad for easy data entry and instrument reading. RS232 port for external communication and Pt100 sensor connection for external monitoring and control. A 4-speed centrifugal pressure / suction pump provides 22-26 L/min flow with 5.8-10.2 psi pressure / 2.9-5.6 psi suction. Integrated programmer with 6 x 60 program steps. Included with each unit: 2 each barbed fittings for tubing 8 and 12 mm inner dia. (pump connections M16x1 female).