Learn More
Biotium CF350 Protein Labeling Kit , 3x(1mg) labelings
Catalog No. 501964922
Encompass_Preferred
This kit contains everything you need to label antibodies or other proteins with a fluorescent CF dye succinimidyl ester (CF dye SE). Each kit contains reagents for three labeling reactions of up to 1 mg protein per reaction. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. UV-excitable blue fluorescent CF350 dye has excitation/emission at 347/448 nm.
Catalog No. 50-196-4922 Supplier Biotium Supplier No. 92210
This kit contains everything you need to label antibodies or other proteins with a fluorescent CF dye succinimidyl ester (CF dye SE). Each kit contains reagents for three labeling reactions of up to 1 mg protein per reaction. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. UV-excitable blue fluorescent CF350 dye has excitation/emission at 347/448 nm.