Learn More
Julabo USA Inc Each, CF40 refrigerated/heating circulator -40…+150 °C
SDP
Catalog No. 507533891
Encompass_Preferred
Bath/Circulators, Julabo, CF40 Compact Refrigerated and Heating Circulator -40...+150 °C. Economy CF30 model is ideal for routine applications with a working temperature range from -40 to +150 °C with a pressure pump. Units feature a splash-proof keypad with LED temperature display, PID temperature control and RS232 interface for PC connection. Included with each unit: 2 each barbed fittings for tubing 8 and 12 mm inner dia. (pump connections M16x1 female).
Catalog No. 50-753-3891 Supplier Julabo USA Inc Supplier No. 9400340.2
Add to Cart
Add to Cart
Request a Quote
Bath/Circulators, Julabo, CF40 Compact Refrigerated and Heating Circulator -40...+150 °C. Economy CF30 model is ideal for routine applications with a working temperature range from -40 to +150 °C with a pressure pump. Units feature a splash-proof keypad with LED temperature display, PID temperature control and RS232 interface for PC connection. Included with each unit: 2 each barbed fittings for tubing 8 and 12 mm inner dia. (pump connections M16x1 female).