Learn More
Biotium CF405L Protein Labeling Kit , 3x(1mg) labelings
Catalog No. 501964932
Encompass_Preferred
This kit contains everything you need to label antibodies or other proteins with a fluorescent CF dye succinimidyl ester (CF dye SE). Each kit contains reagents for three labeling reactions of up to 1 mg protein per reaction. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. 405 nm-excitable green fluorescent CF405L dye has excitation/emission at 395/545 nm.
Catalog No. 50-196-4932 Supplier Biotium Supplier No. 92228
This kit contains everything you need to label antibodies or other proteins with a fluorescent CF dye succinimidyl ester (CF dye SE). Each kit contains reagents for three labeling reactions of up to 1 mg protein per reaction. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. 405 nm-excitable green fluorescent CF405L dye has excitation/emission at 395/545 nm.