Learn More
Biotium CF405L Tyramide
Catalog No. 501964920
Encompass_Preferred
CF dye tyramide conjugates can be used for tyramide signal amplification (TSA), which is an enzyme-mediated detection method that uses horseradish peroxidase (HRP) to generate high-density labeling of a target protein or nucleic acid in situ. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. 405 nm-excitable green fluorescent CF405L dye has excitation/emission at 395/545 nm.Also see Biotium's CF Dye Tyramide Amplification Kits.
Catalog No. 50-196-4920 Supplier Biotium Supplier No. 92198
CF dye tyramide conjugates can be used for tyramide signal amplification (TSA), which is an enzyme-mediated detection method that uses horseradish peroxidase (HRP) to generate high-density labeling of a target protein or nucleic acid in situ. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. 405 nm-excitable green fluorescent CF405L dye has excitation/emission at 395/545 nm.Also see Biotium's CF Dye Tyramide Amplification Kits.