Learn More
Biotium Phalloidin, CF405M
Catalog No. NC0584795
Encompass_Preferred
Phalloidin conjugates are actin filament stains for fixed and permeabilized cells. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Blue fluorescent CF405M has excitation/emission at 408/452 nm.
Catalog No. NC0584795 Supplier Biotium Supplier No. 00034T
Phalloidin conjugates are actin filament stains for fixed and permeabilized cells. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Blue fluorescent CF405M has excitation/emission at 408/452 nm.