Learn More
Biotium CF405S Amine
Catalog No. 501964854
Encompass_Preferred
CF Dye Amines can be used to conjugate CF dyes to carboxylic acids groups in biological molecules. Carboxylic acids can be activated using water soluble carbodiimides to react with amine-functionalized dyes to form a stable amide bond. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Blue fluorescent CF405S dye has excitation/emission at 404/431 nm.
Catalog No. 50-196-4854 Supplier Biotium Supplier No. 92036
CF Dye Amines can be used to conjugate CF dyes to carboxylic acids groups in biological molecules. Carboxylic acids can be activated using water soluble carbodiimides to react with amine-functionalized dyes to form a stable amide bond. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Blue fluorescent CF405S dye has excitation/emission at 404/431 nm.