Learn More
Biotium CF405S Donkey Anti-Rabbit, Highly Cross-Adsorbed, 2mg/mL
Catalog No. 501964346
Encompass_Preferred
Fluorescently labeled Donkey Anti-Rabbit IgG (H+L), Highly Cross-Adsorbed is prepared by labeling high quality antibody with a selection of fluorescent CF dyes and other labels. CF dyes are next-generation fluorescent dyes with advantages brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Blue fluorescent CF405S has excitation/emission at 404/431 nm. Supplied at 2 mg/mL in PBS, 50% glycerol, 2 mg/ml BSA, 0.05% sodium azide.
Catalog No. 50-196-4346 Supplier Biotium Supplier No. 2042050UL
Fluorescently labeled Donkey Anti-Rabbit IgG (H+L), Highly Cross-Adsorbed is prepared by labeling high quality antibody with a selection of fluorescent CF dyes and other labels. CF dyes are next-generation fluorescent dyes with advantages brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Blue fluorescent CF405S has excitation/emission at 404/431 nm. Supplied at 2 mg/mL in PBS, 50% glycerol, 2 mg/ml BSA, 0.05% sodium azide.