Learn More
Biotium CF488A, Aminooxy
Catalog No. 501964862
Encompass_Preferred
Aminooxy dyes have an aminooxy reactive group, which can be used to label a target molecule with an aldehyde or ketone group. An aminooxy group readily reacts with an aldehyde or ketone group to form a stable linkage without the use of a reducing agent. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Green fluorescent CF488A dye has excitation/emission at 490/515 nm.
Catalog No. 50-196-4862 Supplier Biotium Supplier No. 92051
Aminooxy dyes have an aminooxy reactive group, which can be used to label a target molecule with an aldehyde or ketone group. An aminooxy group readily reacts with an aldehyde or ketone group to form a stable linkage without the use of a reducing agent. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Green fluorescent CF488A dye has excitation/emission at 490/515 nm.