Learn More
Biotium Annexin V, CF488A Conjugate, Azide-Free, Lyophilized
Catalog No. 501964411
Encompass_Preferred
CF488A Annexin V is a green fluorescent conjugate (Ex/Em 490/515 nm) that can be used to stain phosphatidylserine on the surface of apoptotic cells. This Annexin V conjugate is supplied lyophilized and azide-free, compatible with real-time imaging of live cells. Unit size 5 ug.
Catalog No. 50-196-4411 Supplier Biotium Supplier No. 29005R5UG
CF488A Annexin V is a green fluorescent conjugate (Ex/Em 490/515 nm) that can be used to stain phosphatidylserine on the surface of apoptotic cells. This Annexin V conjugate is supplied lyophilized and azide-free, compatible with real-time imaging of live cells. Unit size 5 ug.