Learn More
Biotium CF488A-Dextran 10, 000 MW, Anionic and Fixable
Catalog No. 501964790
Encompass_Preferred
CF dye labeled dextrans could be used as a fluorescent fluid-phase markers to study cell permeability, endocytosis, or mechanisms of biomolecular delivery. The dextran conjugates are available in a variety of molecular weights, and contain a formaldehyde-fixable free-amine group. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Green fluorescent CF488A dye has excitation/emission at 490/515 nm.
Catalog No. 50-196-4790 Supplier Biotium Supplier No. 80110
CF dye labeled dextrans could be used as a fluorescent fluid-phase markers to study cell permeability, endocytosis, or mechanisms of biomolecular delivery. The dextran conjugates are available in a variety of molecular weights, and contain a formaldehyde-fixable free-amine group. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Green fluorescent CF488A dye has excitation/emission at 490/515 nm.