Learn More
Biotium CF488A Donkey Anti-Mouse IgG (H+L), Highly Cross-Adsorbed, 2 mg/mL
Supplier:  Biotium 200141
Encompass_Preferred
This product is prepared by labeling highly cross-adsorbed donkey anti-mouse IgG (H+L) with CF488A dye. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Green fluorescent CF488A dye has excitation/emission at 490/515 nm. To minimize cross-reactivity, the antibody has been adsorbed against bovine, chicken, goat, guinea pig, Syrian hamster, horse, human, rabbit, and sheep serum.
Catalog No. NC0995731