Learn More
Biotium CF488A Donkey Anti-Rabbit IgG (H+L), Highly Cross-Adsorbed, 2 mg/mL
Catalog No. NC0995725
Encompass_Preferred
This product is prepared by labeling highly cross-adsorbed donkey anti-rabbit IgG (H+L) with CF488A dye. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Green fluorescent CF488A dye has excitation/emission at 490/515 nm. To minimize cross-reactivity, the antibody has been adsorbed against bovine, chicken, goat, guinea pig, Syrian hamster, horse, human, mouse, rat, and sheep serum.
Catalog No. NC0995725 Supplier Biotium Supplier No. 200151
This product is prepared by labeling highly cross-adsorbed donkey anti-rabbit IgG (H+L) with CF488A dye. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Green fluorescent CF488A dye has excitation/emission at 490/515 nm. To minimize cross-reactivity, the antibody has been adsorbed against bovine, chicken, goat, guinea pig, Syrian hamster, horse, human, mouse, rat, and sheep serum.