Learn More
Biotium CF488A Maleimide
Catalog No. 501964843
Encompass_Preferred
CF Dye Maleimides are thiol-reactive fluorescent dyes. Maleimides are commonly used to label proteins on free thiol groups. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Green fluorescent CF488A dye has excitation/emission at 490/515 nm.
Catalog No. 50-196-4843 Supplier Biotium Supplier No. 92022
CF Dye Maleimides are thiol-reactive fluorescent dyes. Maleimides are commonly used to label proteins on free thiol groups. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Green fluorescent CF488A dye has excitation/emission at 490/515 nm.