Learn More
Biotium CF488A Rabbit Anti-RFP, 1mg/mL
Catalog No. 501964347
Encompass_Preferred
Polyclonal anti-RFP is designed to detect red fluorescent protein (RFP) and its variants: mCherry, tdTomato, mBanana, mOrange, mPlum, and mStrawberry. The antibody was prepared from antiserum to RFP (Discosoma). This product is prepared by labeling the rabbit anti-RFP antibody with CF488A dye. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Green fluorescent CF488A dye has excitation/emission at 490/515 nm.
Catalog No. 50-196-4347 Supplier Biotium Supplier No. 20421
Polyclonal anti-RFP is designed to detect red fluorescent protein (RFP) and its variants: mCherry, tdTomato, mBanana, mOrange, mPlum, and mStrawberry. The antibody was prepared from antiserum to RFP (Discosoma). This product is prepared by labeling the rabbit anti-RFP antibody with CF488A dye. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Green fluorescent CF488A dye has excitation/emission at 490/515 nm.