Learn More
Biotium CF488A TUNEL Assay Apoptosis Detection Kit
Catalog No. 501964490
Encompass_Preferred
In the TUNEL assay, TdT enzyme catalyzes the addition of labeled dUTP to the 3'- ends of the internucleosomal DNA strand breaks that are a hallmark of apoptosis. The CF dye TUNEL Assay Apoptosis Detection Kit employs dUTP conjugated to Biotium's exceptionally bright and photostable CF fluorescent dyes, for single step fluorescent TUNEL labeling of fixed cells or tissues for analysis by fluorescence microscopy or flow cytometry. This kit contains CF488A which has an excitation/emission of 490/515 nm. The kit has reagents sufficient for 50 reactions.
Catalog No. 50-196-4490 Supplier Biotium Supplier No. 30063
In the TUNEL assay, TdT enzyme catalyzes the addition of labeled dUTP to the 3'- ends of the internucleosomal DNA strand breaks that are a hallmark of apoptosis. The CF dye TUNEL Assay Apoptosis Detection Kit employs dUTP conjugated to Biotium's exceptionally bright and photostable CF fluorescent dyes, for single step fluorescent TUNEL labeling of fixed cells or tissues for analysis by fluorescence microscopy or flow cytometry. This kit contains CF488A which has an excitation/emission of 490/515 nm. The kit has reagents sufficient for 50 reactions.