Learn More
Medchemexpress LLC HY-112610 5mg Medchemexpress, CF53 CAS:1808160-52-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1126105MG
Encompass_Preferred
Medchemexpress, HY-112610 5mg CF53 CAS:1808160-52-2 CF53 is a highly potent, selective and orally active inhibitor of BET protein, with a K i of <1 nM, K d of 2.2 nM and an IC 50 of 2 nM for BRD4 BD1. CF53 binds to both the BD1 and BD2 domains of BRD2, BRD3, BRD4, and BRDT BET proteins with high affinities, very selective over non-BET bromodomain-containing proteins. CF53 shows potent anti-tumor activity both in vitro and in vivo [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2427