Learn More
Biotium CF532 Protein Labeling Kit , 3x(1mg) labelings
Supplier:  Biotium 92208
Encompass_Preferred
This kit contains everything you need to label antibodies or other proteins with a fluorescent CF dye succinimidyl ester (CF dye SE). Each kit contains reagents for three labeling reactions of up to 1 mg protein per reaction. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Orange fluorescent CF532 dye has excitation/emission at 527/558 nm.
Catalog No. 50-196-4921