Learn More
Biotium CF543 Donkey Anti-Rabbit IgG (H+L), Highly Cross-Adsorbed, 2 mg/mL
Catalog No. NC0995727
Encompass_Preferred
This product is prepared by labeling highly cross-adsorbed donkey anti-rabbit IgG (H+L) with CF543 dye. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Orange fluorescent CF543 dye has excitation/emission at 541/560 nm. To minimize cross-reactivity, the antibody has been adsorbed against bovine, chicken, goat, guinea pig, Syrian hamster, horse, human, mouse, rat, and sheep serum.
Catalog No. NC0995727 Supplier Biotium Supplier No. 203081
This product is prepared by labeling highly cross-adsorbed donkey anti-rabbit IgG (H+L) with CF543 dye. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Orange fluorescent CF543 dye has excitation/emission at 541/560 nm. To minimize cross-reactivity, the antibody has been adsorbed against bovine, chicken, goat, guinea pig, Syrian hamster, horse, human, mouse, rat, and sheep serum.