Learn More
Biotium Secondary Antibody Human IgG Goat CF555 0 1/EA
Catalog No. 501964273
Encompass_Preferred
This product is prepared by labeling highly cross-adsorbed goat anti-human IgG (H plus L) with CF and reg 555 dye CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes Orange-red fluorescent CF and reg 555 dye has excitation/emission at 555/565 nm To minimize cross-reactivity the antibody has been adsorbed against bovine horse and mouse serum
Catalog No. 50-196-4273 Supplier Biotium Supplier No. 20230
Add to Cart
Edge
Add to Cart
This product is prepared by labeling highly cross-adsorbed goat anti-human IgG (H plus L) with CF and reg 555 dye CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes Orange-red fluorescent CF and reg 555 dye has excitation/emission at 555/565 nm To minimize cross-reactivity the antibody has been adsorbed against bovine horse and mouse serum