Learn More
Biotium CF568 Maleimide
Catalog No. 501964845
Encompass_Preferred
CF Dye Maleimides are thiol-reactive fluorescent dyes. Maleimides are commonly used to label proteins on free thiol groups. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Red fluorescent CF568 dye has excitation/emission at 562/583 nm.
Catalog No. 50-196-4845 Supplier Biotium Supplier No. 92024
CF Dye Maleimides are thiol-reactive fluorescent dyes. Maleimides are commonly used to label proteins on free thiol groups. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Red fluorescent CF568 dye has excitation/emission at 562/583 nm.