Learn More
Biotium Streptavidin, CF568 Conjugate
Supplier:  Biotium 29035
Encompass_Preferred
Streptavidin conjugates are high quality streptavidin biotin binding protein labeled with CF dyes and a selection of other labels. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Red fluorescent CF568 dye has excitation/emission at 562/583 nm.
Catalog No. NC1097512