Learn More
Biotium Annexin V, CF594 Conjugate, Azide-Free, Lyophilized
Supplier:  Biotium 29011R5UG
Encompass_Preferred
CF594 Annexin V is a deep red fluorescent conjugate (Ex/Em 593/614 nm) that can be used to stain phosphatidylserine on the surface of apoptotic cells. This Annexin V conjugate is supplied lyophilized and azide-free, compatible with real-time imaging of live cells. Unit size 5 ug.
Catalog No. 50-196-4415