Learn More
Biotium CF594 Biocytin
Supplier:  Biotium 92006
Encompass_Preferred
Fluorescent biocytin conjugates are cell-impermeant, fixable polar tracers with biotin and an aldehyde-fixable primary amine groups.  Polar tracers are commonly used to investigate cell-cell and cell-liposome fusion as well as membrane permeability and transport through gap junctions or cell uptake during pinocytosis. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Deep-red fluorescent CF594 dye has excitation/emission at 593/614 nm.
Catalog No. 50-196-4838