Learn More
Biotium CF633 Maleimide
Catalog No. 501964846
Encompass_Preferred
CF Dye Maleimides are thiol-reactive fluorescent dyes. Maleimides are commonly used to label proteins on free thiol groups. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Far-red fluorescent CF633 dye has excitation/emission at 630/650 nm.
Catalog No. 50-196-4846 Supplier Biotium Supplier No. 92026
CF Dye Maleimides are thiol-reactive fluorescent dyes. Maleimides are commonly used to label proteins on free thiol groups. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Far-red fluorescent CF633 dye has excitation/emission at 630/650 nm.