Learn More
Biotium CF640R Aminooxy
Catalog No. 501964864
Encompass_Preferred
Aminooxy dyes have an aminooxy reactive group, which can be used to label a target molecule with an aldehyde or ketone group. An aminooxy group readily reacts with an aldehyde or ketone group to form a stable linkage without the use of a reducing agent. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Far-red fluorescent CF640R dye has excitation/emission at 642/662 nm.
Catalog No. 50-196-4864 Supplier Biotium Supplier No. 92058
Aminooxy dyes have an aminooxy reactive group, which can be used to label a target molecule with an aldehyde or ketone group. An aminooxy group readily reacts with an aldehyde or ketone group to form a stable linkage without the use of a reducing agent. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Far-red fluorescent CF640R dye has excitation/emission at 642/662 nm.