Learn More
Biotium CF647 Amine
Supplier:  Biotium 92042
Encompass_Preferred
CF Dye Amines can be used to conjugate CF dyes to carboxylic acids groups in biological molecules. Carboxylic acids can be activated using water soluble carbodiimides to react with amine-functionalized dyes to form a stable amide bond. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Far-red fluorescent CF647 dye has excitation/emission at 650/665 nm.
Catalog No. 50-196-4860