Learn More
Biotium CF647 Goat Anti-Mouse IgG2a (γ2a), 2mg/mL
Supplier:  Biotium 20262
Encompass_Preferred
CF Dye goat anti-mouse IgG2a isotype-specific secondary antibody conjugates are labeled with our exceptionally bright, photostable CF dyes and other labels. The conjugates are prepared from affinity-purified antibodies that react with Fc portion of the heavy chain of mouse IgG2a. To minimize cross reactivity, the antibodies are cross-adsorbed against other mouse IgG subclasses (IgG1, IgG2b, IgG3), and human, bovine and rabbit serum proteins. Far-red fluorescent CF647 has excitation/emission at 650/665 nm. Supplied at 2 mg/mL in PBS, 50% glycerol, 2 mg/ml BSA, 0.05% sodium azide.
Catalog No. 50-196-4288