Learn More
Biotium Phalloidin, CF647
Catalog No. NC1526409
Encompass_Preferred
Phalloidin conjugates are actin filament stains for fixed and permeabilized cells. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Far-red fluorescent CF647 dye has excitation/emission at 650/665 nm.
Catalog No. NC1526409 Supplier Biotium Supplier No. 00041T
Phalloidin conjugates are actin filament stains for fixed and permeabilized cells. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Far-red fluorescent CF647 dye has excitation/emission at 650/665 nm.