Learn More
Biotium CF660C Goat Anti-Rabbit IgG (H+L), Highly Cross-Adsorbed, single label for STORM, 1 mg/mL
Supplier:  Biotium 20813500UL
Encompass_Preferred
This product is prepared by labeling highly cross-adsorbed goat anti-rabbit IgG (H+L) with CF660C dye. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Far-red fluorescent CF660C dye has excitation/emission at 667/685 nm. To minimize cross-reactivity, the antibody has been adsorbed against human, mouse, and rat serum. Single label antibodies have an average of one dye molecule per antibody molecule, which is reported to be optimal for super-resolution imaging by STORM.
Catalog No. 50-196-4375