Learn More
Biotium CF660R Tyramide
Catalog No. 501964919
Encompass_Preferred
CF dye tyramide conjugates can be used for tyramide signal amplification (TSA), which is an enzyme-mediated detection method that uses horseradish peroxidase (HRP) to generate high-density labeling of a target protein or nucleic acid in situ. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Far-red fluorescent CF660R dye has excitation/emission at 663/682 nm.Also see Biotium's CF Dye Tyramide Amplification Kits.
Catalog No. 50-196-4919 Supplier Biotium Supplier No. 92195
CF dye tyramide conjugates can be used for tyramide signal amplification (TSA), which is an enzyme-mediated detection method that uses horseradish peroxidase (HRP) to generate high-density labeling of a target protein or nucleic acid in situ. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Far-red fluorescent CF660R dye has excitation/emission at 663/682 nm.Also see Biotium's CF Dye Tyramide Amplification Kits.