Learn More
Biotium CF680 Goat Anti-Mouse IgG1 (γ1), 2mg/mL
Supplier:  Biotium 20253
Encompass_Preferred
CF dye goat anti-mouse IgG1 isotype-specific secondary antibody conjugates are labeled with our exceptionally bright, photostable CF dyes and other labels. The conjugates are prepared from affinity-purified antibodies that react with Fc portion of the heavy chain of mouse IgG1. To minimize cross reactivity, the antibodies are cross-adsorbed against other mouse IgG subclasses (IgG2a, IgG2b, IgG3), and human, bovine and rabbit serum proteins. Far-red/ near-IR fluorescent CF680 has excitation/emission at 681/698 nm. Supplied at 2 mg/mL in PBS, 50% glycerol, 2 mg/ml BSA, 0.05% sodium azide.
Catalog No. 50-196-4285