Learn More
Biotium CF700 Succinimidyl Ester
Catalog No. 501965007
Encompass_Preferred
CF Dye Succinimidyl Esters (SE, also known as NHS esters) are Biotium's next-generation fluorescent dyes in amine-reactive form for labeling antibodies and other proteins on amine groups, such as lysine residues. Near-infrared CF700 has excitation/emission at 695/720 nm.
Catalog No. 50-196-5007 Supplier Biotium Supplier No. 96067
CF Dye Succinimidyl Esters (SE, also known as NHS esters) are Biotium's next-generation fluorescent dyes in amine-reactive form for labeling antibodies and other proteins on amine groups, such as lysine residues. Near-infrared CF700 has excitation/emission at 695/720 nm.