Learn More
Biotium CF750 Goat Anti-Mouse IgG (H+L)
Supplier:  Biotium 200701MG
Encompass_Preferred
Fluorescently labeled Goat Anti-Mouse IgG (H+L) is prepared by labeling high quality antibody with a selection of fluorescent CF dyes and other labels. CF dyes are next-generation fluorescent dyes with advantages brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Near-IR fluorescent CF750 has excitation/emission at 755/777 nm. Supplied lyophilized, reconstitute with 0.5 ml water to 2 mg/mL in pH ~7.4 PBS containing 15 mg/mL bovine serum albumin (IgG-free and protease-free) and 20 mg/mL trehalose.
Catalog No. 50-196-4182