Learn More
Biotium CF750 Maleimide
Catalog No. 501965005
Encompass_Preferred
CF Dye Maleimides are thiol-reactive fluorescent dyes. Maleimides are commonly used to label proteins on free thiol groups. CF750 (Ex/Em 755/777 nm) is one of Biotium's next-generation near-infrared dyes, with superior brightness, water-solubility, and photostability. Unit size 0.5 umol.
Catalog No. 50-196-5005 Supplier Biotium Supplier No. 96062
CF Dye Maleimides are thiol-reactive fluorescent dyes. Maleimides are commonly used to label proteins on free thiol groups. CF750 (Ex/Em 755/777 nm) is one of Biotium's next-generation near-infrared dyes, with superior brightness, water-solubility, and photostability. Unit size 0.5 umol.