Learn More
Biotium CF770R Succinimidyl Ester
Catalog No. 501964884
Encompass_Preferred
CF Dye Succinimidyl Esters (SE, also known as NHS esters) are amine-reactive fluorescent dyes. SE dyes are commonly used to label antibodies and other proteins on free amine groups, such as lysine residues. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Far-red/near-IR fluorescent CF770R dye has excitation/emission at 770/701 nm.
Catalog No. 50-196-4884 Supplier Biotium Supplier No. 92107
CF Dye Succinimidyl Esters (SE, also known as NHS esters) are amine-reactive fluorescent dyes. SE dyes are commonly used to label antibodies and other proteins on free amine groups, such as lysine residues. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Far-red/near-IR fluorescent CF770R dye has excitation/emission at 770/701 nm.