Learn More
Biotium CF790 Free Acid
Catalog No. 501964907
Encompass_Preferred
Near-IR CF dye free acids are control dyes for studies that involve CF dye succinimidyl ester conjugated to antibody, protein or peptides. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Near-infrared fluorescent CF790 dye has excitation/emission at 784/806 nm.
Catalog No. 50-196-4907 Supplier Biotium Supplier No. 92159
Near-IR CF dye free acids are control dyes for studies that involve CF dye succinimidyl ester conjugated to antibody, protein or peptides. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Near-infrared fluorescent CF790 dye has excitation/emission at 784/806 nm.