Learn More
Biotium CF790 Succinimidyl Ester
Catalog No. 501964903
Encompass_Preferred
CF Dye Succinimidyl Esters (SE, also known as NHS esters) are amine-reactive fluorescent dyes. SE dyes are commonly used to label antibodies and other proteins on free amine groups, such as lysine residues. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Near-infrared fluorescent CF790 dye has excitation/emission at 784/806 nm.
Catalog No. 50-196-4903 Supplier Biotium Supplier No. 92155
CF Dye Succinimidyl Esters (SE, also known as NHS esters) are amine-reactive fluorescent dyes. SE dyes are commonly used to label antibodies and other proteins on free amine groups, such as lysine residues. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Near-infrared fluorescent CF790 dye has excitation/emission at 784/806 nm.