Learn More
Biotium CF820 Succinimidyl Ester
Catalog No. 501965008
Encompass_Preferred
CF Dye Succinimidyl Esters (SE, also known as NHS esters) are Biotium's next-generation fluorescent dyes in amine-reactive form for labeling antibodies and other proteins on amine groups, such as lysine residues. Near-infrared CF820 has excitation/emission at 822/835 nm.
Catalog No. 50-196-5008 Supplier Biotium Supplier No. 96068
CF Dye Succinimidyl Esters (SE, also known as NHS esters) are Biotium's next-generation fluorescent dyes in amine-reactive form for labeling antibodies and other proteins on amine groups, such as lysine residues. Near-infrared CF820 has excitation/emission at 822/835 nm.