Learn More
Chondrex Inc CFA, 1 MG/ML X 5 ML
Catalog No. 501526841
Encompass_Preferred
Heat-killed M. tuberculosis H37 RA (non-viable) suspended in Incomplete Freund's Adjuvant at a concentration of 1 mg/ml. 5 mL.
Catalog No. 50-152-6841 Supplier Chondrex Inc Supplier No. 7008
Heat-killed M. tuberculosis H37 RA (non-viable) suspended in Incomplete Freund's Adjuvant at a concentration of 1 mg/ml. 5 mL.