Learn More
Chondrex Inc CFA, 4 MG/ML X 5 ML
Catalog No. 501526837
Encompass_Preferred
Heat-killed M. tuberculosis H37 RA (non-viable) suspended in Incomplete Freund's Adjuvant at a concentration of 4 mg/ml. 5 mL.
Catalog No. 50-152-6837 Supplier Chondrex Inc Supplier No. 7001
Heat-killed M. tuberculosis H37 RA (non-viable) suspended in Incomplete Freund's Adjuvant at a concentration of 4 mg/ml. 5 mL.