Learn More
Chondrex Inc CFA, 5 MG/ML X 5 ML
Catalog No. 501526844
Encompass_Preferred
Heat-killed M. tuberculosis H37 RA (non-viable) suspended in Incomplete Freund's Adjuvant at a concentration of 5 mg/ml. 5 mL.
Catalog No. 50-152-6844 Supplier Chondrex Inc Supplier No. 7023
Heat-killed M. tuberculosis H37 RA (non-viable) suspended in Incomplete Freund's Adjuvant at a concentration of 5 mg/ml. 5 mL.