Learn More
ABclonal Technology CFAP44 Rabbit pAb
SDP
Supplier:  ABclonal Technology A145391000UL
Encompass_Preferred
Enables peptidase activity. Involved in sperm axoneme assembly. Acts upstream of or within microtubule cytoskeleton organization. Predicted to be located in cytoplasm, cytoskeleton, and motile cilium. Implicated in spermatogenic failure 20.
Catalog No. 50-001-08238