Learn More
Accuflow Systems C-Flex Braided Tubing 25-foot, ID 0.375" x OD 0.625" x Wall 0.125" is fully characterized, validated and tested to a variety of specifications including USP Class VI, ISO 10993 and European Pharmacopoeia.
Catalog No. 501539422
Encompass_Preferred
C-Flex Braided Tubing is an ultra-flexible, high-purity tubing product developed for use in applications where a thermoplastic tube is desired, but the pressure requirements exceed those allowable by a non-reinforced thermoplastic material. C-Flex Braided Tubing is manufactured with C-Flex formula 082, clear, animal derived component free (ADCF) resin and reinforced with polyester braid. C-Flex Braided Tubing is manufactured from the finest grade of thermoplastic materials and is fully characterized, validated and tested to a variety of specifications including USP Class VI, ISO 10993 and European Pharmacopoeia. Available in 25 foot coils for the smaller sizes and 15 foot lengths for the larger sizes, ½" ID through ¾" ID.
Catalog No. 50-153-9422 Supplier Accuflow Systems Supplier No. ACC42029
C-Flex Braided Tubing is an ultra-flexible, high-purity tubing product developed for use in applications where a thermoplastic tube is desired, but the pressure requirements exceed those allowable by a non-reinforced thermoplastic material. C-Flex Braided Tubing is manufactured with C-Flex formula 082, clear, animal derived component free (ADCF) resin and reinforced with polyester braid. C-Flex Braided Tubing is manufactured from the finest grade of thermoplastic materials and is fully characterized, validated and tested to a variety of specifications including USP Class VI, ISO 10993 and European Pharmacopoeia. Available in 25 foot coils for the smaller sizes and 15 foot lengths for the larger sizes, ½" ID through ¾" ID.