Learn More
Aviva Systems Biology Corp CFD Antibody - C-terminal region (ARP60174_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60174_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CFD. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human CFD. Alternate names: ADIPSIN, ADN, DF, PFDApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-286-970