Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ CFD (Human) ELISA Kit

Catalog No. 89101119 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
CCS, Plasma, Serum, Urine
Cell Culture, Milk, Plasma, Saliva, Serum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-101-119 Cell Culture, Milk, Plasma, Saliva, Serum
89-028-427 CCS, Plasma, Serum, Urine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-101-119 Supplier Abnova™ Supplier No. KA3823
Add to Cart
Add to Cart

In-Vitro Enzyme-Linked Immunosorbent assay for the quantitative measurement of Human CFD

Specifications

Conjugate Biotin
Cross Reactivity Human
Description CFD (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human CFD.
Detection Limit 1.8ng/ml
Detection Method Colorimetric
Includes 96 wells-Complement Factor D Microplate, 3 Sealing Tapes, Complement Factor D Standard (480ng), 50X Biotinylated Complement Factor D Antibody (140uL), 10X EIA Diluent Concentrate (30mL), 20X Wash Buffer Concentrate (2 x 30mL), Streptavidin-Peroxidase Conjugate (SP Conjugate) (80uL), Chromogen Substrate (8mL), Stop Solution (12mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Cell Culture, Milk, Plasma, Saliva, Serum
Sample Volume 50 μL
Content And Storage Store the kit at 4°C. For long term storage, please store at -20°C.
For Use With (Application) Quantification
Product Type CFD (Human) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.