Learn More
Apexbio Technology LLC CFDA SE Cell Tracer Kit,1 kit
SDP
Catalog No. 502240036
Encompass_Preferred
The CFDA SE (carboxyfluorescein diacetate succinimidyl ester) Cell Tracer Kit is usually used in fluorescence analyses as a long-term tracer of cells. The Kit can be used for in vitro and in vivo labeling of cells to determine whether or not a cell is proliferating, especially.CFDA SE is a cell-permeable dye, which could bind covalently to all free amines on the surface and inside of cells. It shows little cytotoxicity, with minimal observed effect on the proliferative ability or biology of cells. The intense fluorescent staining provided by CFDA SE which distinguish labeled cells from auto-fluorescence can enable the visualization even after several days post stain, making it an ideal stain for cell tracing studies. Other sizes are also available. Please inqury us for quote.
This item is not returnable. View return policy
Catalog No. 50-224-0036 Supplier Apexbio Technology LLC Supplier No. K10231
The CFDA SE (carboxyfluorescein diacetate succinimidyl ester) Cell Tracer Kit is usually used in fluorescence analyses as a long-term tracer of cells. The Kit can be used for in vitro and in vivo labeling of cells to determine whether or not a cell is proliferating, especially.CFDA SE is a cell-permeable dye, which could bind covalently to all free amines on the surface and inside of cells. It shows little cytotoxicity, with minimal observed effect on the proliferative ability or biology of cells. The intense fluorescent staining provided by CFDA SE which distinguish labeled cells from auto-fluorescence can enable the visualization even after several days post stain, making it an ideal stain for cell tracing studies. Other sizes are also available. Please inqury us for quote.